Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cholesteryl hemisuccinate

Catalog No. C65125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
5 g
25 g

Catalog No. C65125G

Supplier: Sigma Aldrich C65125G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cholesteryl hemisuccinate (CHEMS) is an acidic cholesterol ester. It self assembles into bilayers in alkaline and neutral aqueous media. CHEMS exhibits pH sensitive polymorphism. It consists of succinic acid esterified to the beta-hydroxyl group of cholesterol. This chemical modification results in the ability of CHEMS to adopt a lamellar assembly in suitable aqueous media, whereas cholesterol forms monohydrate crystals in an aqueous environment.

Specifications

CAS 1510-21-0
Molecular Formula C31H50O4
Linear Formula C31H50O4
MDL Number MFCD00037705
Quantity 5 g
Synonym 3 beta-Hydroxy-5-cholestene 3-hemisuccinate; 5-Cholesten-3 beta-ol 3-hemisuccinate; Cholesteryl hydrogen succinate
Molecular Weight (g/mol) 486.73
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.