Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Total AMPK alpha 1 DuoSet IC ELISA

Catalog No. DYC31972
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC31972 2 Plates
DYC31975 5 Plates
DYC3197E 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC31972 Supplier R&D Systems™ Supplier No. DYC31972
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.

ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of IFN-γ/IL-2. Complete ELISpot kits are ready-to-run and require no assay development or refinement.

The Human/Mouse/Rat Total AMPK alpha 1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat AMPK alpha 1.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 625.00 to 40000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 5562
Synonym AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5 '-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5 '-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, PRKAA1, protein kinase, AMP-activated, alpha 1 catalytic subunit
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.