Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Non-sterile Snap Cap for Round Bottom Tubes

Catalog No. FB1495615 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Outer:
12 mm
17 mm
For Use With (Equipment):
14 mL Round Bottom Tube
5 mL Round Bottom Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer For Use With (Equipment)
FB1495615 17 mm 14 mL Round Bottom Tube
FB1496112 12 mm 5 mL Round Bottom Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FB1495615
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fisherbrand™'s snap caps provide reliable containment for various laboratory fluid samples and preparations.

  • Cap only
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
  • Non-sterile
TRUSTED_SUSTAINABILITY

Specifications

Product Type Culture Tube Closure
Diameter (Metric) Outer 17 mm
Description Snap Cap
Material Polyethylene
Color Translucent
Packaging Bulk
Product Line Fisherbrand
Sterility Non-sterile
For Use With (Equipment) 14 mL Round Bottom Tube
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.