Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurofascin Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5144624

Catalog No. FEPA5144624


Only null left
Add to Cart

Description

Description

Neurofascin Polyclonal Antibody for Western Blot

Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neurofascin
Polyclonal
Unconjugated
NFASC
AA387016; D430023G06Rik; DKFZp686P2250; KIAA0756; KIAA0756FLJ46866; mKIAA0756; neurofascin; neurofascin homolog; NF; Nfasc; NRCAML
Rabbit
Antigen affinity chromatography
RUO
116690, 269116
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.8 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300
P97685, Q810U3
NFASC
Recombinant protein encompassing a sequence within the N-terminus region of mouse Neurofascin. The exact sequence is proprietary.
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.