Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Seamless Cellulose Dialysis Tubing
Shop All Fisherbrand Products

Click to view available options
Diameter (Metric):
6.4 mm
15.9 mm
20.4 mm
28.6 mm
32 mm
45 mm
47.7 mm
Length (Metric):
3 m
30 m
Description
- Material: Seamless cellulose
- Molecular Weight Cut-Off (MWCO): Retains molecules ≥12,000
- Da Applications: Ideal for educational settings in analytical chemistry, biology, and pharmaceutical studies
- Permeability: High permeability to water and low molecular weight compounds
- Pore Diameter: Approx. 4.8 nm
- Formulation: Glycerin, water, and 1% sulfur
- Storage: Store in an airtight container; rehumidify if dried out
- Diameter: 45 mm (1.13 in.)
- Length: 30 m (100 ft.)
- Quantity: Sold individually (Each)
- Educational Use Only: No Certificate of Analysis (C of A) provided
This product is for educational use - no C of A is available.
Specifications
Specifications
| Product Type | Dialysis Tubing |
| Material | Seamless Cellulose |
| Diameter (Metric) | 47.7 mm |
| Diameter (English) | 1.88 in. |
| Length (English) | 10 ft. |
| Length (Metric) | 3 m |
| Formulation | Glycerin, water and 1% sulfur |
| Quantity | Each |
| For Use With (Application) | Useful in analytical chemistry, biology, and pharmaceutical fields |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction