Learn More
Carson™ MicroMini 20X LED Pocket Microscope
Shop All Carson Optical Products
Click to view available options
Magnification:
20x
Description
- Attach the digiscoping adapter clip to a smartphone and then use the built-in camera application to take pictures and record videos.
- Lightweight, compact and portable
- Contains a built in LED light to be used in conjunction with the microscope.
- Built-in stand alone LED and UV flashlight to illuminate subject.
- Ideal for educational purposes and can be used for a variety of hobbies, tasks, learning and exploration.
- Excellent for examining plant/blood cells, printing pixels, fabrics, or hair follicles
- Use the stand alone UV flashlight for detecting counterfeit currency, nectar mapping and more.
- For multiple age groups ranging from a young child explorer, middle/high school student to the everyday professional.
- Small enough to fit inside the palm of the hand.
- Compatible with nearly all smartphones including iPhone, Samsung, HTC, LG and more.
- Powered by 3 G12 button cell batteries (included).
- Lifetime Limited Warranty
- Fits all phones
Specifications
Specifications
| Magnification | 20x |
| Product Type | Microscope |
| Lighting | LED |
| Includes | Built-in LED and UV Flashlight and Universal Smartphone Digiscoping Adapter Clip |