Learn More
Carson™ BugView Quick-Release Bug Catching Tool and Magnifier
SDPSCI_ED
Catalog No. S81156 Shop All Carson Optical Products
Click to view available options
Magnification:
5x
Carson™ BugView Quick-Release Bug Catching Tool and Magnifier is a fun and humane way to view the insects, nature and wildlife. Temporarily trap the specimen, view the bug and then release it back into its environment. Its thumb operated trapping slider makes it easy to catch the fastest specimens in a safe and humane fashion.
  • Temporarily trap the specimen, view the bug and then release it back into its environment.
  • Thumb-operated trapping slider makes it easy to catch the fastest specimens in a safe and humane fashion.
  • Contains Built-in 5X Power acrylic magnifier for examining intricate details on subjects
  • Excellent educational tool for children of all ages to learn and explore the environment around them.
  • Can also be used by adults for trapping and releasing bugs and spiders around the home or office in hard to reach places without harming them
  • Limited Lifetime Warranty
Specifications
Product Type Magnifier
Magnification 5x
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.