Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC)

Catalog No. L32478
Encompass_Preferred
Click to view available options
Quantity:
1 mg

Bright-green LEL-fluorophore conjugate

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC), is a bright-green LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-FITC can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

• Very bright FITC conjugate detected with traditional FITC filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Tomato
Conjugate Fluorescein
Content And Storage When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Label Type Classic Dyes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.