Learn More
CNPase Mouse anti-Bovine, Canine, Human, Mouse, Pig, Rat, Rabbit, Sheep, Clone: 11-5B, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB326R
Description
This Anti-CNPase Antibody, clone 11-5B is validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of CNPase.
Specifications
CNPase | |
Monoclonal | |
Unconjugated | |
NM_033133.4 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C for one year | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
11-5B | |
Purified mouse monoclonal IgG1 in buffer containing 0.02M Phosphate buffer, pH 7.6, 0.25M NaCl with 0.1% sodium azide. | |
CNP, CNPase, CNP1, EC 3.1.4.37 | |
Purified human 2’, 3’-cyclic nucleotide 3’-phosphodiesterase | |
100 μg | |
Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat, Sheep | |