Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Decarboxylase Mouse anti-Human, Rat, Clone: GAD-6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB351
Description
Anti-GAD Antibody. 65 kDa isoform, clone GAD-6 detects level of Glutamate Decarboxylase and has been published and validated for use in immunohistochemistry and western blotting.
Specifications
Glutamate Decarboxylase | |
Monoclonal | |
Unconjugated | |
NM_000818.1 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
Immunohistochemistry, Western Blot | |
GAD-6 | |
Purified immunoglobulin. Lyophilized from 10mM potassium phosphate, 70mM sodium chloride, pH 7.4, 0.02% sodium azide. Reconstitute with 100μL of sterile water (1mg/mL). | |
GAD2, MGC161607, GAD65, MGC161605, GAD-65, EC 4.1.1.15 | |
Purified rat brain glutamic acid decarboxylase. | |
100 μg | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction