Learn More
Epredia™ Lab Vision™ 14.3.3σ Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 1433S01
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human 14.3.3σ protein
Molecular Weight: 26-30kDa
Positive Control: HT29 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | 14.3.3sigma Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1433S01 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human 1433 sigma protein |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.