Learn More
Description
Host Species: Mouse
Clone: MLuC5
Isotype: IgM
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Small cell lung carcinoma N592 live cells
Molecular Weight: 67kDa
Positive Control: Breast CA
Cellular Localization: Membrane with some cytoplasmic Ab-1 partially inhibits the binding of soluble laminin.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
- Inhibits (partially) the binding of soluble laminin
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Laminin Receptor Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MLuC5 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Small cell lung carcinoma N592 live cells |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Research Discipline | Extracellular Matrix |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.