Learn More
Description
Host Species: Mouse
Clone: 1B12
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant external domain of CD23 protein
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD23 Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1B12 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant external domain of CD23 protein |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Oncogenesis |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.