Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX17 Antibody (SR2137), Novus Biologicals™
SDP

Catalog No. NB260193 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260193 50 μg
NB258623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260193 Supplier Novus Biologicals Supplier No. NBP32215650UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DDX17 Monoclonal antibody specifically detects DDX17 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen DDX17
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR2137
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 17, DEAD box protein 17, DEAD box protein p72, DEAD/H box 17, DKFZp761H2016, EC 3.6.1, EC 3.6.4.13, P72DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD), probable ATP-dependent RNA helicase DDX17, RH70, RNA-dependent helicase p72
Host Species Rabbit
Immunogen A synthesized peptide derived from human DDX17 (Uniprot #: Q92841)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Translation Control, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10521
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.