Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB266442 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB266442 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB266442 Supplier Novus Biologicals Supplier No. NBP222328JF549
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TXNDC Polyclonal antibody specifically detects TXNDC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TXNDC
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp564E1962, PDIA11, protein disulfide isomerase family A, member 11, thioredoxin domain containing 1, thioredoxin domain-containing, Thioredoxin domain-containing protein 1, thioredoxin-related transmembrane protein 1, TMXTXNDC, Transmembrane Trx-related protein, TXNDC1thioredoxin domain containing
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 230 to 280 of human Thioredoxin-Related Transmembrane Protein 1 using the numbering given in entry NP_110382.3 (GeneID 81542).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 81542
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.