Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscle Phosphofructokinase/PFKM/PFK-1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB267539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB267539 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB267539 Supplier Novus Biologicals Supplier No. NBP232170JF549
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Muscle Phosphofructokinase/PFKM/PFK-1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 730 to 780 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5213
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.