Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ High Sample Diluents: B-Reagent

Catalog No. OCD8321200
Change view
Click to view available options
Quantity:
1 x 28 mL + 100 Wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD8321200 1 x 28 mL + 100 Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD8321200 Supplier QuidelOrtho Supplier No. 8321200
Add to Cart
Add to Cart

Specifications

Chemical Name or Material High sample diluent B pack (Buffered)
For Use With (Equipment) Vitros™ Myoglobin assay
Quantity 1 x 28 mL + 100 Wells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.