Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529049 Supplier Invitrogen™ Supplier No. PA529049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HL-60 membrane extracts. Predicted reactivity: Mouse (95%), Rat (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Guanylyl cyclases, catalyzing the production of cGMP from GTP, are classified as soluble and membrane forms. The membrane guanylyl cyclases, often termed guanylyl cyclases A through F, form a family of cell-surface receptors with a similar topographic structure: an extracellular ligand-binding domain, a single membrane-spanning domain, and an intracellular region that contains a protein kinase-like domain and a cyclase catalytic domain. GC-A and GC-B function as receptors for natriuretic peptides; they are also referred to as atrial natriuretic peptide receptor A and type B. Also see NPR3, which encodes a protein with only the ligand-binding transmembrane and 37-amino acid cytoplasmic domains. NPR1 is a membrane-bound guanylate cyclase that serves as the receptor for both atrial and brain natriuretic peptides and BNP, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPR1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NPR1
Gene Accession No. P16066, P18910
Gene Alias AI893888; ANPa; ANP-A; ANPRA; ANPR-A; atrial natriuretic peptide A-type receptor; atrial natriuretic peptide receptor 1; atrial natriuretic peptide receptor A; atrial natriuretic peptide receptor type A; atrionatriuretic peptide receptor A; Gca; GC-A; Guanylate cyclase A; guanylyl cyclase-A; GUC2A; GUCY2A; natriuretic peptide A type receptor; natriuretic peptide receptor 1; natriuretic peptide receptor A; natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptide receptor A); Nppa receptor; Npr1; Npra; NPR-A; Pndr; testicular tissue protein Li 20
Gene Symbols NPR1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human NPR-A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100343663, 24603, 4881
Target Species Human, Rabbit, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.