Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579275
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579275 Supplier Invitrogen™ Supplier No. PA579275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. IHC: mouse testis tissue, rat testis tissue.

Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Fmn1
Gene Accession No. Q05860, Q68DA7
Gene Alias FMN; Fmn1; formin (limb deformity); formin 1; formin-1; Ld; limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100
Gene Symbols Fmn1
Host Species Rabbit
Immunogen E. coli-derived human FMN1 recombinant protein (Position: N1195-N1419).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14260, 296512, 342184
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.