Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARTS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582767 Supplier Invitrogen™ Supplier No. PA582767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDDDLEFRPP SRPQSSDNQQ YFCAPAPLSP SARPRSPWGK LDPYDSSEDD KEYVGFATLP NQVHRKS Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 84%.

Apoptosis is related to many diseases and development. Mitochondrial proteins, such as cytochrome c, Apaf-1, and AIF play important role in apoptosis. A novel mitochondrial septin-like protein was identified recently and designated ARTS for apoptosis related protein in TGF-beta signaling pathway. ARTS that is encoded by the human septin H5/PNUTL2/CDCrel2b gene is located to mitochondria and translocates to the nucleus when apoptosis occurs. ARTS is expressed in many tissues. It enhances cell death induced by TGF-beta and, to a lesser extent, by other apoptotic agents, such as TNF-a and Fas ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARTS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPTIN4
Gene Accession No. O43236
Gene Alias 4-Sep; apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bh5; BRADEION; Bradeion beta; brain protein H5; C17orf47; CE5B3; CE5B3 beta; cell division control-related protein 2; cell division control-related protein 2b; cerebral protein 7; EG3-1RVC; EG3RVC; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; H5; hCDCREL-2; hucep-7; MART; M-Septin; OTTMUSG00000001265; peanut-like 2; peanut-like 2 homolog; peanut-like protein 2; PNUTL2; SEP4; Sept4; septin 4; septin H5; Septin4; septin-4; septin-M; Uncharacterized protein C17orf47
Gene Symbols SEPTIN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEPT4. Recombinant protein control fragment (Product #RP-93189).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.