Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPBB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592950 Supplier Thermo Scientific Supplier No. PA592950
Add to Cart
Add to Cart

GPBB Polyclonal Antibody

GPBB Polyclonal Antibody for Western Blot, ICC/IF, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPBB
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PYGB
Gene Accession No. P11216, P53534, Q8CI94
Gene Alias brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB
Gene Symbols PYGB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 544-843 of human PYGB (NP_0028532).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110078, 25739, 5834
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.