Learn More
Thermo Scientific™ MUG Disk
Catalog No. R21135
Click to view available options
Quantity:
25 Disks/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Reagent-impregnated disk recommended for use in rapid, presumptive identification of Escherichia coli.
Presumptively identify Escherichia coli by fluorogenic assay procedure using Thermo Scientific™ Remel™ MUG Disk. In 1976, Kilian and Bulow reported the enzyme β-glucuronidase is present in most strains of E. coli (97%)1. The substrate, 4-methylumbelliferyl- β-D-glucuronide (MUG), is found to be both sensitive and selective for detection of β-glucuronidase activity. Trepeta and Edberg used the MUG test in conjunction with oxidase, indole, and lactose fermentation for rapid, cost effective identification of E. coli2.
Godsey et al. supported their findings in an evaluation of methods based on microbial enzyme activity profiles3. Most strains of E. coli produce an enzyme β-D-glucuronidase. In the indole test, the enzyme tryptophanase attacks tryptophan molecule on its side chain, leaving the aromatic ring in the form of indole. The indole is then detected by addition of ρ-dimethylamino-benzaldehyde (Kavocs’ Reagent) which produces a red color. β-D-glucuronidase is an enzyme that cleaves the substrate 4-methylumbelliferyl-β-D-glucuronide (MUG) and thereby produces a fluorescent product called methylumbelliferone4. This compound is detectable using a long wave ultraviolet light (360 nm).
  • Ready to use - 4-methylumbelliferyl- β-D-glucuronide-impregnated disk
  • Easy to store - No special storage required
  • Easy to interpret - With visual inspection
    • In MUG Disk test, examine the disk for fluorescence using a longwave ultraviolet light (360nm) in a darkened room. Blue fluorescence is interpreted as positive test.
    • In Indole test, add one drop of Kovacs‘ Indole Reagent to the disk and immediately observe for red color development.
Interpretation:
MUG Disk Test (Direct or Tube Test):
Positive test - Blue fluorescence observed under 360 nm UV light
Negative test - No fluorescence observed under 360 nm UV light
Indole Test:
Positive test - Red color development on the disk or in the tube
Negative test - No red color development on the disk or in the tube
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes E. coli
Type E. coli Testing Kit
Quantity 25 Disks/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.