Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Periodic Table of the Elements, with Pictures of Elements
SDPSCI_ED

Catalog No. s20743 Shop All 3B Scientific Products
Click to view available options
Quantity:
1

3B Scientific™ Periodic Table of the Elements, with Pictures of Elements is displayed on strong laminated material with rods and a hanging cord

Specifications

Includes Poster
Width (English) 78.6 in.
Height (English) 54.3 in.
Quantity 1
Material Plastic
Color White
Languages English, German, French, Spanish
Subject Chemistry
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.