Learn More
3B Scientific™ Gynecologic Skills Trainer P91
GREENER_CHOICESDPSCI_ED
Catalog No. s27320 Shop All 3B Scientific Products
3B Scientific™ Gynecologic Skills Trainer P91 is an adult-sized lower torso designed as a versatile clinical training tool to add hands-on experience for learners practicing gynecologic skills.
Trainer will be useful for demonstrating the following procedures:
  • Bi-manual examinations with palpable, realistic, normal and abnormal uteri
  • Vaginal examination with speculum
  • Train vaginal examination with easily interchangeable cervices and have visual recognition of normal and abnormal anatomy variations
  • Evaluate multiple conditions using the interchangeable normal and pathologic cervices
  • Pap test for cervical screening
  • Practice catheterization (no fluid)
  • Suppository administration and digital rectal examination
Additional Features:
  • SKINlike™ high-quality silicone offers a realistic look, feel and texture in addition to a life-like softness.
  • Four different uteri modules with externally palpable abnormal pathologies are easily interchangeable and can be completed with 6 different cervices.
  • Adult-sized female lower torso with accurate anatomy, including ischial spines, pubic bones and sacrum for a realistic examination
  • Leg stumps already in examination position and with the gynecologic examination wedge the possibility to simulate a leaned position of the patient as in a gynecological examination chair
  • Bladder can be simulated full or empty to make the palpation of the structure possibly restricted
  • Offers realistic flexibility of the vaginal canal wall
  • Seamless, flexible skin for practice of speculum placement
  • Labia can be realistically separated
  • Urethra built for catheterization simulation with CH 14 catheter
  • Abdominal wall can be removed quickly and without any tools to change the uterus and the cervix easily
  • Latex free
Includes:
  • Gynecologic Skills Trainer P91
  • Uteri set (4):
    • Normal uterus
    • Bicornate uterus
    • Uterus with 3 removable myomas structures
    • Enlarged uterus
  • Cervices Set (6):
    • Normal nulliparous or para 0 cervix
    • Normal multiparous cervix
    • Ectropion cervix (erosion)
    • Dysplasia cervix
    • Cervix with carcinoma
    • Cervix with barrel-shaped carcinoma
  • Gynecologic examination wedge
  • Lubricant (2 x 250 mL)
Specifications
For Use With (Application) Gynecological Skills Training
Includes Gynecologic Skills Trainer P91, Uteri set (4), Cervices Set (6), Examination wedge, Lubricant
Material SKINlike™ High-quality Silicone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.