Learn More
3B Scientific™ Buttock Injection Simulator
GREENER_CHOICESDPSCI_ED
Catalog No. s27327 Shop All 3B Scientific Products
Click to view available options
:
buttock injection simulator
3B Scientific™ Buttock Injection Simulator is a high-quality, life-like strap-on intramuscular simulator of a right buttock
  • Simulator features anatomical landmarks for intramuscular (I.M.) injections:
    • Iliac crest
    • Anterior superior iliac spine
    • Greater trochanter
  • Integrated fine electronics provide graphic feedback when training the intramuscular injection technique, and correctly administered injections will produce audiovisual feedback
  • Additional training and control modes are available for checking results either immediately or upon completion of a training session.
  • Includes a 21G/0.8 injection needle and 5 mL syringe.
Specifications
Product Type Buttock Injection Simulator
Includes 21G/0.8 injection needle and 5 mL syringe
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.