Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Using CRISPR to Treat Cystic Fibrosis Kit
SDPSCI_ED

Catalog No. s27552 Shop All EDVOTEK Products
Click to view available options
Class Size:
8 Lab Groups

Simulate the use of CRISPR-Cas9 to target a genetic mutation found in a patient suffering from Cystic Fibrosis. Students will develop an understanding of guide RNA (gRNA) design, and use agarose gel electrophoresis to examine pre-prepared DNA samples after CRISPR treatment.

  • Storage: Room Temperature Stable
  • Refrigerator Storage of Ready-to-Load QuickStrip™ samples is recommended
  • Suitable for 8 Gels
  • Expected Time: 45 minutes

Specifications

Product Type Kit
Grade 9 to Undergraduate
Age 14 to 20
Includes Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50 X), 10 x Gel Loading Solution, FlashBlue™ DNA Stain
Requires DNA Electrophoresis, Micropipettes: 5-50 μL (Optional), White Light Box, and Microwave/Hot Plate
Time Range 45 min.
Content And Storage Room Temperature Stable, Refrigerate Ready-to-Load QuickStrip™ samples
Class Size 8 Lab Groups
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.