Learn More
ZOLL™ Medical Electrodes for AED Plus™ Defibrillators
SCI_ED
Catalog No. s38753
Click to view available options
Product Type:
Electrode, CPR-D-padz
Electrode, Pedi-padz II
This item is not returnable. View return policy
This item is not returnable. View return policy
For complete CPR and defibrillation delivery
  • ZOLL’s intelligent pediatric capability is built into the AED Plus. The AED can sense when Pedi-padz II are being used and will switch the AED Plus from adult to pediatric mode, modifying the device so it will provide an appropriate pediatric ECG analysis algorithm and therapy energy level.
  • For use with patients under the age of 8 years or <55 pounds
  • Two-year shelf life
Specifications
Product Type Electrode, Pedi-padz II
Size Pediatric
For Use With (Equipment) AED Plus Defibrillator
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.