Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Forensic Drug Testing: A Simulated Immunoassay
Shop All Aldon Products

Click to view available options
Class Size:
15 Lab Groups
Description
- Drug testing comes in two forms: Presumptive and Confirmatory.
- Because of the cost associated with confirmatory testing, a presumptive test is often employed first.
- While there are various forms of presumptive drug testing, two of the most common techniques are immunoassay testing and thin-layer chromatography.
- All urine samples and testing reagents are simulated to avoid the potential hazards of actual biological specimens while still providing realistic results.
- Kit contains enough materials for 15 groups.
- Teacher Guide and Student Study Guide copymasters are included.
- Aligned to the Next Generation Science Standards (NGSS).
Specifications
Specifications
Certifications/Compliance | Next Gen Aligned |
Product Type | Forensic Drug Detection Kit |
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | Forensics |
Includes | 5 mL Positive Control; 5 mL Negative Control; 5 mL Specimen 1; 5 mL Specimen 2; 5 mL Specimen 3; 5 mL Specimen 4; 5 mL Specimen 5; 5 mL Specimen 6; 30 mL Antibody Solution; 30 mL Labeled Metabolite; 15 Spot Plates |
Material | Cardboard Box Kit |
Quantity | 1 |
Dimensions (L x W x H) | 9 x 6.5 x 2.8 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction