Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Potassium SMART 3 Reagent System
SDPSCI_ED

Catalog No. s95107
Click to view available options
No. of Tests:
100

Potassium reagent system for use with SMART 3 Colorimeter.

  • Range 0.8 to 10.0 ppm
  • 100 tests
  • Tetraphenylboron Method

Specifications

pH Range 6.8 to 8.4
For Use With (Application) For Industrial Waters
For Use With (Equipment) SMART 3 Colorimeter
No. of Tests 100
Range 0.8 to 10.0 ppm
Test Method Tetraphenylboron Method
Type Reagent System
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.