Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Phosphate Buffered Saline (PBS)

Catalog No. SH30256FS Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
Concentration:
1 X
10 X
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
SH30256FS 6 x 500 mL 1 X
SH3025602 1000 mL 1 X
SH3025601 500 mL 1 X
SH30256LS 6 x 1000 mL 1 X
SH3025801 500 mL 10 X
SH3025802 1000 mL 10 X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SH30256FS Supplier Cytiva Supplier No. SH30256.FS
Add to Cart
Add to Cart

Buffer cell culture growth with PBS

  • QC-tested solution
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Phosphate Buffered Saline (PBS)
Sterilization Method 0.1 μm Sterile-filtered
Without Additives No Calcium, No Magnesium, No Phenol Red
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.2
Quantity 6 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.