Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ DAB Quanto Detection System

Catalog No. ta999qhdx Shop All Epredia Products
Click to view available options
Quantity:
1 L
125 mL DAB Quanto Substrate, 4 mL DAB Quanto Chromagen
60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.

Provides a strong brown staining and can be used after reconstitution for up to 1 week at room temperature and up to 2 weeks at 2° to 8°C.

DAB Quanto has been validated in combination with both UltraVision LP and UltraVision Quanto and provides four major benefits.
    • Strong staining—ideal to quantifying cells and protein expression
    • Less mixing—reducing personnel time and stress
    • Less disposal—cuts down waste
    • Lower risk of mixing errors—supports higher staining consistency
    3,3' Diaminobenzidine (DAB) is a widely used chromogen for immunohistochemical staining and immunoblotting. The DAB Quanto is an entirely new formula for a diaminobenzidine-based peroxidase substrate that develops faster on tissues sections reducing incubation times. It utilizes novel chemistry to provide greater sensitivity, generating a crisper, darker, insoluble precipitate. The standard working dilution is 30μL of DAB Quanto Chromogen per 1ml of DAB Quanto Substrate. The use of liquid components reduces some risks associated with the handling of powders or tablets. The DAB Quanto offers an added advantage, as the working solution is stable for up to 2 weeks at 2 to 8°C and up to a week at room temperature enabling continued use and minimizing waste disposal.
    • Supplied in a two-component form consisting of a DAB Quanto Chromogen and DAB Quanto Substrate
    • Standard working dilution is 30μL of DAB Quanto Chromogen per 1mL of DAB Quanto Substrate
    • Use of liquid components reduces some risks associated with the handling of powders or tablets

    DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.
  • Specifications

    Chromagen Type DAB Quanto Chromagen
    Quantity 1 L
    Substrate Type Dab Quanto Subrate
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.