Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 1070722
GREENER_CHOICE

Catalog No. 443110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-311-0 10 mg
44-315-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-311-0 Supplier Tocris Bioscience™ Supplier No. 4431/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent, selective GSK-3 inhibitor

Potent glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 0.6 nM for GSK-3α and GSK-3β). Displays high selectivity (> 50-fold) for GSK-3 over a panel of other kinases tested, including CDK family members. Decreases phosphorylation of microtubule-associated protein Tau in vitro; protects rat primary cortical neurons against β amyloid and glutamate challenge. Brain penetrant.

Chemical Identifiers

CAS 1384424-80-9
Molecular Formula C17H13F3N4O2

Specifications

Quantity 10 mg
Formula Weight 362.31
Purity >99%
Molecular Weight (g/mol) 362.312
Chemical Name or Material A 1070722
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.