Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 1899
SDP

Catalog No. 69725 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-725 5 mg
69-722-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-725 Supplier Tocris Bioscience™ Supplier No. 6972/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent K2P3.1 (TASK-1) and K2P9.1 (TASK-3) blocker

A 1899 is a potent K2P3.1 (TASK-1) and K2P9.1 (TASK-3) channel blocker (IC50 values are 7 nM and 70 nM for human TASK-1 and TASK-3 expressed in CHO cells, respectively). Displays >12.5-fold selectivity for TASK-1 and TASK-3 over other K+ channels (IC50 = 0.9 μM for K2P18.1/TRESK and >2 μM for all other channels tested). Binds within open channel pore.

Specifications

Chemical Name or Material N-[(2,4-Difluorophenyl)methyl]-2'-[[[2-(4-methoxyphenyl)acetyl]amino]methyl][1,1'-biphenyl]-2-carboxamide
CAS 498577-46-1
Quantity 5 mg
Molecular Formula C30H26F2N2O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.