Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 196
GREENER_CHOICE

Catalog No. 616710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-671-0 10 mg
61-675-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-671-0 Supplier Tocris Bioscience™ Supplier No. 6167/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective SUV420H1/H2 inhibitor

A 196 is a potent and selective SUV420H1/H2 inhibitor (IC50 values are 25 and 144 nM respectively) that exhibits 100-fold selectivity over other histone methyltransferases and non-epigenetic targets. A 196 inhibits the di- and tri-methylation of H4K20me in multiple cell lines (IC50 < 1 μM).

To request the negative control for A 196, please fill out the SGC 2043 request form on the SGC website.

Specifications

Chemical Name or Material 6,7-Dichloro-N-cyclopentyl-4-(4-pyridinyl)-1-phthalazinamine
CAS 1982372-88-2
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C18H16Cl2N4
Formula Weight Observed MW: 359.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.