Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 205804
GREENER_CHOICESDP

Catalog No. 252450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-245-0 50 mg
25-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-245-0 Supplier R&D Systems™ Supplier No. 2524/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of E-selectin and ICAM-1 expression

Selective inhibitor of E-selectin and ICAM-1 expression (IC50 values are 20, 25 and > 1000 nM for TNF-α-induced E-selectin, ICAM-1 and VCAM-1 expression respectively). Effective inhibitor of cell-cell adhesion in an in vitro flow experiment.

Chemical Identifiers

CAS 251992-66-2
Molecular Formula C15H12N2OS2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 300.39
Purity >98%
Molecular Weight (g/mol) 300.394
Chemical Name or Material A 205804
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.