Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 412997 dihydrochloride
GREENER_CHOICE

Catalog No. 455225
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-522-5 25 mg
45-525 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-522-5 Supplier Tocris Bioscience™ Supplier No. 4552/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective D4 agonist

A 412997 dihydrochloride is a selective agonist for the dopamine D4 receptor (Ki values are 7.9 and 12.1 nM for human D4 and rat D4 receptors). Displays no affinity (<1000 nM) for other dopamine receptors. Shows efficacy in rat models of ADHD and short-term memory.

Specifications

Chemical Name or Material N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
CAS 1347744-96-0
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C19H23N3O.2HCl
Formula Weight Observed MW: 382.33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.