Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 425619
GREENER_CHOICE

Catalog No. 578110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-811-0 10 mg
57-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-811-0 Supplier Tocris Bioscience™ Supplier No. 5781/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TRPV1 antagonist

A 425619 is a potent TRPV1 antagonist (IC50 = 3-9 nM). Blocks capsaicin-evoked currents in rat DRG neurons and inhibits TRPV1-activation by anandamide (Cat. No. 1339) and N-arachidonoyl-dopamine. Reduces pain in inflammatory, postoperative and osteoarthritic in vivo pain models.

Specifications

Chemical Name or Material N-5-Isoquinolinyl-N′-[[(4-(trifluoromethyl)phenyl]methyl]urea
CAS 581809-67-8
Quantity 10 mg
Target TRPV Antagonists
Molecular Formula C18H14F3N3O
Formula Weight 345.32
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.