Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 438079 hydrochloride
GREENER_CHOICE

Catalog No. 297210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-7210 10 mg
29-725-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-7210 Supplier Tocris Bioscience™ Supplier No. 2972/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Competitive P2X7 antagonist

A 438079 hydrochloride is a competitive P2X7 receptor antagonist (pIC50 = 6.9 for the inhibition of Ca2+ influx in the human recombinant P2X7 cell line). Devoid of activity at other P2 receptors (IC50 > 10 μM). Possesses antinociceptive activity in models of neuropathic pain in vivo. Inhibits methamphetamine-induced microglial migration and phagocytosis in vitro.

Chemical Identifiers

CAS 899431-18-6
Molecular Formula C13H10Cl3N5

Specifications

Quantity 10 mg
Formula Weight 342.61
Purity >98%
Molecular Weight (g/mol) 342.608
Chemical Name or Material A 438079 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.