Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 484954
GREENER_CHOICESDP

Catalog No. 448350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-835-0 50 mg
44-831-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-835-0 Supplier Tocris Bioscience™ Supplier No. 4483/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaM kinase III (eEF-2 kinase) inhibitor

Inhibitor of eukaryotic elongation factor-2 (eEF-2) kinase (IC50 = 0.28 μM in enzymatic assay). Reduces eEF-2 phosphorylation with little effect on cancer cell growth.

Chemical Identifiers

CAS 142557-61-7
Molecular Formula C13H15N5O3

Specifications

Quantity 50 mg
Formula Weight 289.29
Purity >98%
Molecular Weight (g/mol) 289.295
Chemical Name or Material A 484954
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.