Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 485
GREENER_CHOICESDP

Catalog No. 63875 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-875 5 mg
63-872-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-875 Supplier Tocris Bioscience™ Supplier No. 6387/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Histone Acetyltransferase Inhibitors

A 485 is a potent and selective p300/CREB-binding protein (CBP) HAT domain inhibitor (IC50 values are 2.6 and 9.8 nM for the CBP-bromodomain HAT-C/H3 (BHC) and p300-BHC domains, respectively), which displays > 1000-fold selectivity over closely related HATs. A 485 suppresses proliferation in several hematological malignancies and AR+ prostate cancer cell lines in vitro, and also inhibits tumor growth in a castration-resistant prostate cancer xenograft model. A 485 is orally bioavailable.

To request the negative control for A 485, please fill out the A 486 request form on the SGC website.

Specifications

CAS 1889279-16-6
Synonym (1R)-N-[(4-Fluorophenyl)methyl]-2,3-dihydro-5-[[(methylamino)carbonyl]amino]-2′,4′-dioxo-N-[(1S)-2,2,2-trifluoro-1-methylethyl]spiro[1H-indene-1,5′-oxazolidine]-3′-acetamide
Quantity 5 mg
Molecular Formula C25H24F4N4O5
Formula Weight 536.48
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.