Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 582941
GREENER_CHOICESDP

Catalog No. 434110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-411-0 10 mg
43-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-411-0 Supplier Tocris Bioscience™ Supplier No. 4341/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Partial agonist at α7 nAChR

Selective α7 nAChR partial agonist; exhibits high affinity for both rat and human α7 receptors (Ki values are 10.8 and 16.7 nM respectively). Displays ∽15-fold selectivity for α7 over 5-HT3 receptors (Ki = 150 nM for 5-HT3).

Chemical Identifiers

CAS 848591-90-2
Molecular Formula C17H20N4

Specifications

Quantity 10 mg
Formula Weight 280.37
Purity >98%
Molecular Weight (g/mol) 280.375
Chemical Name or Material A 582941
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.