Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 740003
GREENER_CHOICE

Catalog No. 370110R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-011-0R 10 mg
37-015-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-011-0R Supplier Tocris Bioscience™ Supplier No. 3701/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective P2X7 antagonist

A 740003 is a potent and selective P2X7 receptor antagonist (IC50 values are 18 and 40 nM for rat and human receptors respectively). Displays selectivity over a variety of P2X and P2Y receptors up to a concentration of 100 μM. Reduces nociception in animal models of persistent neuropathic and inflammatory pain. Also reduces neuroblastoma tumor growth in mice.

Specifications

Quantity 10 mg
Product Type A 740003
Purity >98%
Molecular Weight (g/mol) 474.55
Inhibitors A 740003

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.