Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 769662
GREENER_CHOICESDP

Catalog No. 333650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-365-0 50 mg
33-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-365-0 Supplier R&D Systems™ Supplier No. 3336/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent AMPK activator

Potent, reversible AMP-activated protein kinase (AMPK) activator (EC50 = 0.8 μM) that displays selectivity towards β1 subunit-containing heterotrimers. Inhibits fatty acid synthesis (IC50 = 3.2 μM) and decreases plasma glucose and triglyceride levels in vivo. Also inhibits proliferation of mesenchymal stem cells, and impedes reprogramming of mouse embryonic fibroblasts to iPSCs. Activates autophagy.

Chemical Identifiers

CAS 844499-71-4
Molecular Formula C20H12N2O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 360.39
Purity >98%
Molecular Weight (g/mol) 360.387
Chemical Name or Material A 769662
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.