Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 77636 hydrochloride
GREENER_CHOICESDP

Catalog No. 170150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-015-0 50 mg
17-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-015-0 Supplier R&D Systems™ Supplier No. 1701/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective D1-like agonist; orally active

Potent and selective dopamine D1-like receptor agonist (pEC50 values are 8.97 and < 5 for D1-like and D2-like receptors respectively). Displays anti-Parkinsonian activity following oral administration in vivo. Exhibits 11-fold cell type bias over dopamine in a functional assay in U2 cells.

Chemical Identifiers

CAS 145307-34-2
Molecular Formula C20H28ClNO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 365.9
Purity >98%
Molecular Weight (g/mol) 365.898
Chemical Name or Material A 77636 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.