Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 784168
GREENER_CHOICESDP

Catalog No. 431950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-195-0 50 mg
43-191-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-195-0 Supplier R&D Systems™ Supplier No. 4319/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective TRPV1 antagonist

Potent TRPV1 antagonist (IC50 = 25 nM for inhibition of TRPV1 activation by 50 nM capsaicin). Displays no activity against a range of receptors, including TRPA1, GABA, opioid, and purinergic receptors.

Chemical Identifiers

CAS 824982-41-4
Molecular Formula C19H15F6N3O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 479.4
Purity >99%
Molecular Weight (g/mol) 479.397
Chemical Name or Material A 784168
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.