Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 939572

Catalog No. 484510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-451-0 10 mg
48-455-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-451-0 Supplier Tocris Bioscience™ Supplier No. 4845/10
This item is not returnable. View return policy

Potent SCD-1 inhibitor

A 939572 is a potent stearoyl-CoA desaturase 1 (SCD1) inhibitor (IC50 = 0.4 nM). Selective for SCD1 over a range of kinases and hERG channels. Inhibits proliferation of squamous cell carcinoma FaDu cells in vitro. Also induces cell death of undifferentiated human embryonic stem cells (ESCs). Orally bioavailable.

Specifications

Chemical Name or Material 4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide
CAS 1032229-3-6
Quantity 10 mg
Target Compounds for Stem Cell Differentiation
Molecular Formula C20H22ClN3O3
Formula Weight 387.86
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.