Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A. aeolicus BPL/BioID2 Antibody (SS QD1), DyLight 405, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP259940V

Catalog No. NBP2-59940V


Only null left
Add to Cart

Description

Description

A. aeolicus BPL/BioID2 Monoclonal specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

A. aeolicus BPL/BioID2
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
GST fused to A. aeolicus BPL/BioID2
0.1 mL
Primary
Bacteria
IgG2 κ
Western Blot, Immunocytochemistry, Immunofluorescence
SS QD1
Western Blot, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
Epitope Tags
This antibody is reactive with A. aeolicus and has not been tested with other species. The birA Entrez Gene number is NP_213397. UniProt number is O66837.
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.