Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A. aeolicus BPL/BioID2 Antibody (SS QD1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259940IR
Description
A. aeolicus BPL/BioID2 Monoclonal specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
A. aeolicus BPL/BioID2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GST fused to A. aeolicus BPL/BioID2 | |
0.1 mL | |
Primary | |
Bacteria | |
IgG2 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
SS QD1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
Epitope Tags | |
This antibody is reactive with A. aeolicus and has not been tested with other species. The birA Entrez Gene number is NP_213397. UniProt number is O66837. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction