Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ alpha-Bungarotoxin

Click to view available options
Quantity:
1 mg
Description
α-Bungarotoxin is a neurotoxin that blocks neuromuscular transmission via irreversible inhibition of nicotinic ACh receptors (nAChRs). Prevents opening of nicotinic receptor-associated ion channels and is selective for α7 receptors over α3β4 receptors (IC50 values are 1.6 nM and > 3 μM respectively).
Specifications
Specifications
| Content And Storage | Store at -20°C |
| Description | a-Bungarotoxin |
| Molecular Weight (g/mol) | 7984.14 |
| Product Type | α-Bungarotoxin |
| Quantity | 1 mg |
| Sequence | Ile-Val-Cys-His-Thr-Thr-Ala-Thr-Ser-Pro-Ile-Ser-Ala-Val-Thr-Cys-Pro-Pro-Gly-Glu-Asn-Leu-Cys-Tyr-Arg-Lys-Met-Trp-Cys-Asp-Ala-Phe-Cys-Ser-Ser-Arg-Gly-Lys-Val-Val-Glu-Leu-Gly-Cys-Ala-Ala-Thr-Cys-Pro-Ser-Lys-Lys-Pro-Tyr-Glu-Glu-Val-Thr-Cys-Cys-Ser-Thr-Asp-Lys-Cys-Asn-Pro-His-Pro-Lys-Gln-Arg-Pro-Gly (Disulfide bridges between 3 - 23, 16 - 44, 29 - 33, 48 - 59, 60 - 65) |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction