Learn More
A Closer Look at Pond Life Lab Investigation
SCI_ED
Catalog No. p-2372515
Click to view available options
Includes:
Pond life lab investigation kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
  • Dried pond mix
  • Clear pond containers
  • Protist-slowing agent
  • Test vials
  • Pipets
  • Vital stain
  • Microscope slides
  • Simulated pollutants
  • Comprehensive teacher and student guides
  • Illustrated, dichotomous key to microlife forms
Observe microbial populations and learn to calculate population diversity indices.
  • Investigate a unique pond ecosystem
  • Build a miniature pond ecosystem
  • Explore the diverse life forms that evolve
  • Monitor microbial populations
  • Gain experience in calculating population diversity indices
  • Design unique experiments to determine the population dynamics within a micro-ecosystem as well as the effects of various pollutants
Order Info
Class size: 40 students.
Specifications
Includes Pond life lab investigation kit
Product Type A Closer Look at Pond Life Lab Investigation
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.